missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Iscove's Modification of DMEM

Catalog No. MT10016CM
Change view
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
With Additives:
Glutamine, HEPES
HEPES
Without Additives:
No α-Thioglycerol, No beta-Mercaptoethanol
No Glutamine, No beta-Mercaptoethanol
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives
MT10016CM 6 x 1 L Glutamine, HEPES No α-Thioglycerol, No beta-Mercaptoethanol
MT10016CV 6 x 500 mL Glutamine, HEPES No α-Thioglycerol, No beta-Mercaptoethanol
MT15016CV 6 x 500 mL HEPES No Glutamine, No beta-Mercaptoethanol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MT10016CM Supplier Corning™ Supplier No. 10016CM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Corning™ Iscove's Modification of DMEM is a highly enriched synthetic medium optimized for rapidly proliferation cultures of high cell density.

Substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over the original formulation. Iscove's Modification of DMEM (IMDM) is a highly enriched synthetic medium optimized for rapidly proliferation cultures of high cell density. A modified form of Dulbecco's Modified eagle Medium (DMEM), Iscove's contains additional amino acids, vitamins, selenium and HEPES buffer, and is formulated with potassium nitrate in place of ferric nitrate. The addition of albumin and transferrin may be required for certain cell types. This formulation is produced with L-glutamine and 25 mM HEPES, and do not contain alpha-thioglycerol and beta-mercaptoethanol.

Specifications

Concentration 1 X
Form Liquid
Product Type Iscove's Modification of DMEM media
Sterility Sterile
With Additives Glutamine, HEPES
Without Additives No α-Thioglycerol, No beta-Mercaptoethanol
Quantity 6 x 1 L
pH 7 to 7.4
Content And Storage 2° to 8°C