missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Hank's Balanced Salt Solution, 10X

Catalog No. mt20023cv
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT20023CV 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT20023CV Supplier Corning™ Supplier No. 20023CV
Only null left

Buffer for maintaining physiological pH of cells maintained in non-CO2 atmospheric conditions

(This is suggested replacement for 20-023-CVR.)

Technical Advantage:  Lower documented Endotoxin content than offerings from other major suppliers
Hanks' Balanced Salt Solution (HBSS) is a buffer used to maintain a physiological pH for cells maintained in non-CO2 atmospheric conditions.
-  Complete Certificate of Analysis available for each production lot, along with full formulation information
-  Produced under the highest industry standards to ensure superior results

Corning media certificates are available on our Resource Library.

Specifications

Content And Storage 2° to 30°C
Product Type Hanks' Balanced Salt Solution
Sterility Sterile
Without Additives No Phenol Red, No Sodium Bicarbonate
pH 4.5 to 5
Quantity 500 mL