missing translation for 'onlineSavingsMsg'
Learn More

Corning™ G418 Sulfate, 50mg/mL

Catalog No. MT30234CR
Change view
Click to view available options
Quantity:
20 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT30234CR 20 mL
MT30234CI 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT30234CR Supplier Corning™ Supplier No. 30234CR
Only null left

Blocks polypeptide synthesis by hindering protein elongation. Corning™ G418 Sulfate is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.

Well-known, effective selection agent G418 Sulfate is recommended for use as selection agent in a concentration range of 100-800 μg/mL. This antibiotic blocks polypeptide synthesis by hindering protein elongation, and inhibits the selection and maintenance of eukaryotic cells stably transfected with the neomycin resistance gene.  G418 is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.

Specifications

Content And Storage 2° to 8°C
Concentration 50 mg/ mL Solution (active G418)
Form Liquid
Product Type Antibiotic
Sterility Sterile
Format Plastic Bottle
Quantity 20 mL