missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Dulbecco's Modification of Eagle's

Catalog No. MT10090CM
Change view
Click to view available options
Quantity:
6 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT10090CM 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT10090CM Supplier Corning™ Supplier No. 10090CM

With low documented Endotoxin content

(This is suggested replacement for 10-090-CMR.)

[+] L-glutamine Technical Advantage:  Lower documented Endotoxin content than offerings from other major suppliers
This 1:1 mixture of Dulbecco's Modified Eagle Medium and Ham's F-12 media is ideal for culture of a variety of cell types under low serum conditions.  Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME.  Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids.  This formulation of DMEM/Ham's F-12 contains L-glutamine.
-  Complete Certificate of Analysis available for each production lot, along with full formulation information
-  Produced under the highest industry standards to ensure superior results The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Corning media certificates are available on our Resource Library.

Specifications

Product Type Dulbecco's Modified Eagle Medium
Quantity 6 x 1 L
Content And Storage 2° to 8°C