missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DMEM With 4.5g/L Glucose, Without L-Glutamine and Sodium Pyruvate

Catalog No. MT15017CM
Change view
Click to view available options
Quantity:
6 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT15017CM 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT15017CM Supplier Corning™ Supplier No. 15017CM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Corning™ DMEM With 4.5g/L Glucose, Without L-Glutamine and Sodium Pyruvate is hybridoma tested.

Growth Promotion and Cytotoxicity

Corning classical media products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Product performance is measured by harvest-to-plant ratios or fold increase according to predetermined specifications.


Physiochemical Properties
Liquid and powder classical media products are tested to determine pH and osmolality. Classical media powders are also tested for residual moisture. The pH and osmolality tests are performed according to specific procedures in which all equipment is calibrated using standards traceable to the National Institute of Standards and Technology.


Biological Tests


Quality release testing of Corning liquid classical media products assures low levels of endotoxin, mycoplasma, and an absence of contamination.


Endotoxin: Classical media products are tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) chromogenic assay. For release, each lot of standard liquid classical media must have an endotoxin level of <0.25 EU/mL. Classical media powder products must have an endotoxin level of <0.25 EU/mL when reconstituted to formula weight with low endotoxin water.


Sterility: All Corning liquid classical media products are tested for sterility.


Mycoplasma: Liquid classical media products are tested for mycoplasma using a large-volume method.

Specifications

Concentration 1 X
Form Liquid
Product Type DMEM Media
With Additives Glucose
Without Additives No Glutamine, No Sodium Pyruvate
Quantity 6 x 1 L
pH 7 to 7.4
Content And Storage 2° to 8°C