missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DMEM With 4.5g/L glucose and sodium pyruvate without L-glutamine

Catalog No. MT15013CV
Change view
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT15013CV 6 x 500 mL
MT15013CM 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT15013CV Supplier Corning™ Supplier No. 15013CV

Substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Corning™ DMEM with L-Glutamine, 4.5 g/L Glucose and Sodium Pyruvate is hybridoma tested.

Growth Promotion and Cytotoxicity

Corning classical media products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Product performance is measured by harvest-to-plant ratios or fold increase according to predetermined specifications.


Physiochemical Properties
Liquid and powder classical media products are tested to determine pH and osmolality. Classical media powders are also tested for residual moisture. The pH and osmolality tests are performed according to specific procedures in which all equipment is calibrated using standards traceable to the National Institute of Standards and Technology.


Biological Tests


Quality release testing of Corning liquid classical media products assures low levels of endotoxin, mycoplasma, and an absence of contamination.


Endotoxin: Classical media products are tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) chromogenic assay. For release, each lot of standard liquid classical media must have an endotoxin level of <0.25 EU/mL. Classical media powder products must have an endotoxin level of <0.25 EU/mL when reconstituted to formula weight with low endotoxin water.


Sterility: All Corning liquid classical media products are tested for sterility.


Mycoplasma: Liquid classical media products are tested for mycoplasma using a large-volume method.

Specifications

Form Liquid
Product Type DMEM Media
With Additives Glucose, Sodium Pyruvate
Without Additives No Glutamine
Quantity 6 x 500 mL
pH 7 to 7.4
Content And Storage 2° to 8°C