Learn More
Corning™ Collection Bags
Collection bags with three ports used for biopharmaceutical applications
Supplier:  Corning™ 9120045
 View more versions of this product
Catalog No. MT9120045
Only null left
Description
Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Specifications
Chemical Identifiers
Heat sealable flexible tubing
1/Ea.
1/Pk.
Each
Specifications
Heat sealable flexible tubing
1/Ea.
Port 1 - 0.37 in. Connector, 12 L x 0.37 I.D. x 0.62 in. O.D., chemically resistant, heat sealable flexible tubing, port 2 - male MPC connector with 0.37 in. barb, female MPC cap, port- 3 female MPC connector with 0.37 in. barb, male MPC plug, break-off i
10 L
EVA
1/Pk.
Each
Product Suggestions
Product Suggestions