Learn More
Corning™ Collection Bags
Catalog No. MT9120042
Change view
Click to view available options
Capacity (Metric):
0.5 L
1 L
5 L
10 L
20 L
50 L
200 L
Film Description:
EVA
ULDPE
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Film Description
MT9120042 1 L EVA
MT9120041 0.5 L EVA
MT9120043 5 L EVA
MT9120047 20 L EVA
MT9120045 10 L EVA
MT9120048 50 L EVA
MT9120083 200 L ULDPE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MT9120042 Supplier Corning™ Supplier No. 9120042
Collection bags with three ports used for biopharmaceutical applications
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Specifications
Tubing Style Heat sealable flexible tubing
Film Description EVA
No. per Case 1/Ea.
No. per Pack 1/Pk.
Port Detail Male MPC connector with 0.25 in. barb, female MPC cap, female MPC connector with 0.25 in. barb, male MPC plug, break-off injection site
Quantity Each
Capacity (Metric) 1 L