Learn More
Corning™ Collection Bags
Collection bags with three ports used for biopharmaceutical applications
Supplier: Corning™ 9120041
Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Chemical Identifiers
| Heat sealable flexible tubing | |
| 1/Ea. | |
| 1/Pk. | |
| Each |
Specifications
| Heat sealable flexible tubing | |
| 1/Ea. | |
| Male MPC connector with 0.25 in. barb, female MPC cap, female MPC connector with 0.25 in. barb, male MPC plug, break-off injection site | |
| 0.5 L |
| EVA | |
| 1/Pk. | |
| Each |