missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Ciprofloxacin Hydrochloride

Catalog No. p-4877223
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT61277RG 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT61277RG Supplier Corning™ Supplier No. 61277RG
Only null left

Packaged in plastic bottles

Specifications

Content And Storage 15° to 30°C
Form Powder
Product Type Antibiotic
Format Plastic Bottle
Quantity 5 g