missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell Culture Media - Dulbecco's Modification of Eagle's Medium (DMEM)

Catalog No. MT10014CV
missing translation for 'changeView'
Click to view available options
Formulation:
With L-Glutamine, 1g/L Glucose and Sodium Pyruvate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
MT10014CV With L-Glutamine, 1g/L Glucose and Sodium Pyruvate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. MT10014CV Supplier Corning™ missing translation for 'supplierNo' 10014CV

Substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over the original formulation. Corning™ Cell Culture Media - Dulbecco's Modification of Eagle's Medium (DMEM) is hybridoma tested.

Growth Promotion and Cytotoxicity

Corning classical media products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Product performance is measured by harvest-to-plant ratios or fold increase according to predetermined specifications.


Physiochemical Properties
Liquid and powder classical media products are tested to determine pH and osmolality. Classical media powders are also tested for residual moisture. The pH and osmolality tests are performed according to specific procedures in which all equipment is calibrated using standards traceable to the National Institute of Standards and Technology.


Biological Tests


Quality release testing of Corning liquid classical media products assures low levels of endotoxin, mycoplasma, and an absence of contamination.


Endotoxin: Classical media products are tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) chromogenic assay. For release, each lot of standard liquid classical media must have an endotoxin level of <0.25 EU/mL. Classical media powder products must have an endotoxin level of <0.25 EU/mL when reconstituted to formula weight with low endotoxin water.


Sterility: All Corning liquid classical media products are tested for sterility.


Mycoplasma: Liquid classical media products are tested for mycoplasma using a large-volume method.

Specifications

Form Liquid
Product Type DMEM Media
Quantity 6 x 500 mL
Formulation With L-Glutamine, 1g/L Glucose and Sodium Pyruvate
Content And Storage 2° to 8°C