Learn More
Corning™ CellCube™ Tubing Adaptor
A new closed system offering designed to help reduce the risk of adventitious contamination
$1073.29 - $4130.25
Specifications
Diameter (Metric) Inner Tubing 9.53 mm
No. per Case 4/Cs.
No. per Pack 1/Pk.
View More Specs
Products 3
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
10-320-190
View Documents
Corning™
3236
4 AseptiQuik™ S Connectors, 4 Tubes Clamps
Case for $4,130.25
Only null left
10-320-191
View Documents
Corning™
3237
AseptiQuik™ S Connected to a Female MPC
Case for $1,073.29
Only null left
10-320-192
View Documents
Corning™
3238
AseptiQuik™ S Connected to a Male MPC
Case for $1,073.29
Only null left
Description
Description
  • Closed System Cell Cube
  • CellCube 10, 25, 100 modules, circulation loops, and connectors
  • Allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik™ and MPC connectors.
Specifications
Specifications
9.53 mm
4/Cs.
1/Pk.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'