Learn More
Corning™ Black Phenolic Threaded Screw Caps
GREENER_CHOICE
Catalog No. 1496230G
Change view
Click to view available options
Neck Finish:
15-415
18-415
38-430
40-400
No. per Pack:
12/Pk.
288/Pk.
32/Pk.
72/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Pack
1496230G 15-415 288/Pk.
028839D 40-400 72/Pk.
028839J 18-415 32/Pk.
028839L 38-430 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 1496230G Supplier Corning™ Supplier No. 9999152
Only null left
Resist effects of temperature and steam in autoclaving
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
Specifications
Color Black
Material Phenolic Resin
No. per Pack 288/Pk.
Liner Material Rubber
Neck Finish 15-415
Height (Metric) 16 mm
Product Type Screw Cap