Learn More
Corning™ Black Phenolic Threaded Screw Caps
GREENER_CHOICE
Resist effects of temperature and steam in autoclaving
Supplier:  Corning™ 9999182
 View more versions of this product
Catalog No. 028839J
Only null left
Description
Description
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
Specifications
Chemical Identifiers
Black
Phenolic Resin
Rubber
Screw Cap
Specifications
Black
32/Pk.
18-415
Screw Cap
Phenolic Resin
Rubber
27 mm
Autoclavable
Product Suggestions
Product Suggestions