Learn More
Corning™ BioCoat™ Matrigel™ 96-Well, Matrigel-Treated, Flat-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Supplier: Corning™ 354607
Description
Corning BioCoat products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.
Specifications
| Standard | |
| 8 x 12 | |
| 96 | |
| Matrigel | |
| For culture of polarized cells and angiogenesis assays, promotes capillary tubule formation of endothelial cells and neurite regeneration |
| With Lid | |
| Clear | |
| Sterile | |
| Flat Bottom (F-well) | |
| 5/Cs. |