missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ 96-Well, Fibronectin-Treated, Flat-Bottom Microplate

Catalog No. 15100162
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15100162 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15100162 Supplier Corning™ Supplier No. 4584
Only null left

Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:Without Lid

High optical quality, glass bottom, black microplates are ideal for performing high content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.

  • High optical quality and scratch resistant glass
  • Glass bottom thickness of 200 μm is well suited for imaging microscopy.
  • Bottom flatness <50 μm to ensure planarity for imaging devices
  • Low background fluorescence and minimal cross-talk provides the highest possible optical quality for cell-based assays.
  • Half area 96-well microplate reduces reagent consumption.

Order Info

Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates

Specifications

Format Half-Area
Lid Without Lid
Content And Storage 2°C to 8°C
Culture Area 0.32 cm2
Array 8 x 12
Color Black, Clear Bottom
No. of Wells 96
Sterility Non-sterile
Surface Treatment Fibronectin
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, increased proliferation of PC12 cells, studies of effects of collagen IV on cell behavior, cell adhesion assays
No. per Case 10/Cs.
No. per Pack 1/Pk.
Source Human Fibronectin
Well Color Clear
Show More Show Less