missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap
GREENER_CHOICE

Catalog No. 08774339
Change view
Click to view available options
Capacity (Metric):
70 mL
250 mL
600 mL
750 mL
Culture Area:
150 cm2
175 cm2
25 cm2
75 cm2
No. per Case:
10/Cs.
40/Cs.
5/Cs.
50/Cs.
Neck Style:
Canted
Straight
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Culture Area No. per Case Neck Style
08774339 750 mL 175 cm2 40/Cs. Straight
08774329 600 mL 150 cm2 5/Cs. Canted
08774328 250 mL 75 cm2 5/Cs. Canted
08774327 70 mL 25 cm2 10/Cs. Canted
08774330 750 mL 175 cm2 5/Cs. Straight
08774336 70 mL 25 cm2 50/Cs. Canted
08774338 600 mL 150 cm2 40/Cs. Canted
08774337 250 mL 75 cm2 50/Cs. Canted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 08774339 Supplier Corning™ Supplier No. 356487
Only null left

Pre-coated with Collagen I for improved cell attachment and an increased proliferation rate of transfected mammalian cells, primary cells and stem cells. Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap provides highly consistent and biologically functional precoated surfaces to more closely mimic in vivo environments for cell culture applications.

Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture

Source

  • Rat tail tendon
  • Quality Control

    • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
    • Tested and found negative for bacteria and fungi
    • Collagen I purity >90% by SDS-PAGE

    Storage

  • 4°C to 30°C under dry conditions.
  • Specifications

    Barcode Yes
    Capacity (English) 25.3 oz.
    Closure Material Phenolic
    Capacity (Metric) 750 mL
    For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
    Neck Style Straight
    No. per Case 40/Cs.
    No. per Pack 5/Pk.
    Shape Rectangular
    Surface Coating Collagen I
    Sterility Sterile
    Bottom Shape Rectangular
    Closure Type Vented
    Culture Area 175 cm2
    Show More Show Less