missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Basic Fibroblast Growth Factors (bFGF), Human Recombinant
GREENER_CHOICE

Catalog No. p-96972
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CB40060 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CB40060 Supplier Corning™ Supplier No. 354060
Only null left

Isolated from pituitary glands

The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.

  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions

Growth factors are proteins that bind to receptors found on the surface of non-hematopoietic cells. Each family of growth factors affect specific cell types, e.g., nerve growth factors (NGF) affect nerve cell types, and epidermal growth factors (EGF) affect epithelial cell types.

Corning offers a range of growth factors that are derived from different species, both natural and recombinant. These growth factors have been used to promote long-term proliferation in cell culture and to induce cell differentiation in a variety of cell types under serum-free or reduced serum conditions.

Order Info

  • FGF is supplied lyophilized, bFGF in 3.6mM Tris, PBS pH 7 buffer
  • bFGF (human) is lyophilized

Specifications

Content And Storage -20°C
Product Type Growth Factor
Formulation Lyophilized from phosphate buffered saline, carrier free, reconstitute in dH2O or serum-free medium
For Use With (Application) Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro
Purity or Quality Grade >95% by SDS-PAGE
Quantity 10 μg
Reactivity Human
Source E. coli