missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Pipette Tips for Molecular Devices FLIPR Tetra Workstation
GREENER_CHOICE

Catalog No. p-5234483
Change view
Click to view available options
Color:
Black
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14223556 Clear
14223557 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14223556 Supplier Axygen™ Supplier No. FT384CR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pipette tips manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality

  • 384 Tip format
  • Polypropylene
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Compatibility: Molecular Devices FLIPR Tetraworkstation
  • Specifications

    Volume (Metric) 30 μL
    Sterility Non-sterile
    Color Clear
    Format Rack
    Tip Style Automation, Round
    For Use With (Equipment) Molecular Devices FLIPR Tetra Automation System, Hamilton MICROLAB STAR
    Material Polypropylene