missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well White Flat Bottom Polystyrene NBS Microplate Without Lid
GREENER_CHOICE

Catalog No. 07201202
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07201202 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07201202 Supplier Corning™ Supplier No. 3990
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nonionic hydrophilic surface

  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • Specifications

    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color White
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    No. per Case 25/Cs.
    No. per Pack 5/Pk.
    Quantity 25/Cs.
    Show More Show Less