Learn More
Corning™ 96-Well, Ultra-Low Binding, U-Shaped-Bottom Microplate
GREENER_CHOICE
Catalog No. 15100173
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
15100173 Sterile
1 options
Catalog No. 15100173 Supplier Corning™ Supplier No. 4520
Color:Clear; Sterility:Sterile; Lid:With Lid
With their novel and proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment (ULA) surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization.
  • Unique design shields each well to minimize well-to-well cross-talk.
  • You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate.
Specifications
Format Standard
Lid With Lid
Array 8 x 12
Packaging 10/Pk., 50/Cs.
Binding Type Ultra-Low Attachment
Color Clear
No. of Wells 96
Sterility Sterile
Surface Treatment Ultra-low Binding
Well Shape Round Bottom (U-well)
No. per Case 50/Cs.
No. per Pack 10/Pk.
Show More Show Less