missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Special Optics Low Fluorescence Assay Plates
GREENER_CHOICE

Catalog No. 07200730
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07200730 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07200730 Supplier Corning™ Supplier No. C3615
Get Up to $2,500 in Products with Purchase
View all promotions

Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • Specifications

    Lid Without Lid
    Array 8 x 12
    Packaging 4 inner packs of 25
    Color Black, Clear Bottom
    Product Type 96-well Assay Microplate (Untreated)
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Non-sterile
    Surface Treatment Non-treated
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Spectroscopy, polarization and imaging
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less