missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Half Area Black Flat Bottom Polystyrene NBS™ Microplate
GREENER_CHOICE

Catalog No. 07201205
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07201205 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07201205 Supplier Corning™ Supplier No. 3993
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to reduce well-to-well crosstalk

  • Designed to reduce well-to-well cross-talk
  • Black microplates have low background fluorescence and minimize light scattering.
  • Specifications

    Format Half-Area
    Lid Without Lid
    Array 8 x 12
    Color Black
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 25 to 125 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    No. per Case 25/Cs.
    No. per Pack 5/Pk.
    Quantity 25/Cs.
    Show More Show Less