missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Clear Polypropylene Corner Notch Microplates
GREENER_CHOICE

Catalog No. 05539200
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
05539200 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05539200 Supplier Corning™ Supplier No. 3879
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured from solvent resistant polypropylene which provides compatibility with many common organic solvents. Corning™ 96-Well Clear Polypropylene Corner Notch Microplates are ideal for growing cells in stationary suspension or where reduced cell attachment is desired.

(This is the suggested replacement for 3790)

  • Designed for growing cells in stationary suspension or other applications where reduced cell attachment is desired
  • Round bottom wells with 330μL total volume
  • Recommended working volumes of 75 to 200μL
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g. DMSO, ethanol, methanol)
  • Nonreversable lids with condensation rings to reduce contamination
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Individual alphanumeric codes for well identification
  • Individually wrapped

Specifications

Lid With Lid
Array 8 x 12
Binding Type Medium Binding
Color Clear
Material Polypropylene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
No. per Case 50/Cs.
No. per Pack 1/Pk.
Quantity 50/Cs.
Show More Show Less