missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Clear Microplate with Generic Barcode
GREENER_CHOICE

Catalog No. 15100128
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15100128 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15100128 Supplier Corning™ Supplier No. 9018BC

Ideal for immunoassays

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

Specifications

Format Standard
Lid Without Lid
Array 8 x 12
Binding Type High Bind
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Non-sterile
Surface Treatment High Binding
Well Shape Flat Bottom (F-well)
For Use With (Application) Ideal for immunoassays
No. per Case 100/Cs.
No. per Pack 25/Pk.
Show More Show Less