missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Non-Binding, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE

Catalog No. 07200844
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07200844 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07200844 Supplier Corning™ Supplier No. C3881
Only null left

Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:Without Lid

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • Specifications

    Format Half-Area
    Lid Without Lid
    Array 8 x 12
    Packaging 25/Pk., 100/Cs.
    Color Black, Clear Bottom
    Product Type Half Area
    For Use With (Equipment) Standard automated handling and detection systems
    Material Polystyrene
    Volume (Metric) Well 190 μL
    No. of Wells 96
    Volume (Metric) Working 25 to 125 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less