missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well, Non-Treated, Flat-Bottom Microplate

Description
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Culture Area | 0.05 cm2 |
| Array | 16 x 24 |
| Color | Black |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 5 to 40 μL |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Show More |