missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well Clear Polystyrene Microplates

Catalog No. 361014632
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
361014632 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361014632 Supplier Corning™ Supplier No. 3640

Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 364-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.

  • Total well volume of 112 μL; working well volume of 20 μL – 80 μL
  • Cell culture microplates are sterile and nonpyrogenic.
  • The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties.
  • Specifications

    Format Standard
    Lid Without Lid
    Description Nonbinding (NBS™) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions
    Array 16 x 24
    Color Clear
    Product Type 384-Well NBS™ Microplate
    Material Polystyrene
    No. of Wells 384
    Volume (Metric) Working 20 to 80 μL
    Sterility Non-sterile
    Surface Treatment Non-treated
    Total Volume (Metric) Well 112 μL
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Well Color Clear
    Show More Show Less