missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well Low-Volume Solid Microplates

Description
Polystyrene plates feature a unique square-to-round well design with a conical-shaped bottom that provides easier access and imaging for superior assay performance.
- Bottom reduces fluid and air entrapment, greater signal-to-noise ratio and reduced FP signal (mP) when working with very low volumes
- Each well has a raised rim to reduce cross-contamination during assay and liquid-handling process
- Choose black plates for fluorescence applications or white for luminescence applications
- Ideal for immunoassays
- Can bind medium (>10kDa) and large biomolecules that possess ionic groups and/or hydrophobic regions
- Working volume: 2 to 20μL
Specifications
Specifications
| Barcode | No Barcode |
| Format | Standard |
| Array | 16 x 24 |
| Color | White |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 5 to 40 μL |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Well Shape | Flat Bottom (F-well) |
| Show More |