missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom, Low Volume Microplate

Description
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.
Specifications
Specifications
| Barcode | No Barcode |
| Format | Standard |
| Lid | With Lid |
| Culture Area | 0.03 cm2 |
| Array | 16 x 24 |
| Binding Type | TC-Treated |
| Color | White |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 5 to 40 μL |
| Show More |