missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 07201320
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07201320 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07201320 Supplier Corning™ Supplier No. C3570
Only null left

Color:White; Sterility:Sterile; Lid:With Lid

 

  • Minimal well to well crosstalk
  • White microplates enhance luminecent signals and have low background luminescence
  • Black mircoplates have low background fluorescence and minimize light scattering
  • Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
  • Medical-grade polystyrene
  • Well volume: 112μL
  • Working volume: 20μL to 80μL
  • Surface area/well: 0.06cm2
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Plate dimensions meet proposed industry standards

 


Specifications

Format Standard
Lid With Lid
Array 16 x 24
Packaging 10/Pk., 50/Cs.
Color White
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
For Use With (Application) Attachment and growth of anchorage-dependent cells
Includes Lid
Quantity 50/Cs.
Show More Show Less