Learn More
Corning™ 384-Well Solid Black or White Polystyrene Microplates
GREENER_CHOICE
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Supplier:  Corning™ 4512
 View more versions of this product
Catalog No. 15100157
Only null left
Description
Description
  • Minimal well to well crosstalk
  • White microplates enhance luminecent signals and have low background luminescence
  • Black mircoplates have low background fluorescence and minimize light scattering
  • Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
  • Medical-grade polystyrene
  • Well volume: 112μL
  • Working volume: 20μL to 80μL
  • Surface area/well: 0.06cm2
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Plate dimensions meet proposed industry standards
Specifications
Chemical Identifiers
Standard
Polystyrene
Non-sterile
16 x 24
384
Specifications
Standard
16 x 24
Polystyrene
5 to 40 μL
Non-treated
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
Without Lid
White
384
Non-sterile
Round Bottom (U-well)
Product Suggestions
Product Suggestions