missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. 07200852
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07200852 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07200852 Supplier Corning™ Supplier No. C3571
Only null left

Color:Black; Sterility:Sterile; Lid:With Lid

Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.

Specifications

Format Standard
Lid With Lid
Culture Area 0.06 cm2
Array 16 x 24
Color Black
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 20 to 80 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
For Use With (Application) Attachment and growth of anchorage-dependent cells
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Show More Show Less