missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom, High Content Microplate
GREENER_CHOICE

Catalog No. 13701103
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
13701103 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13701103 Supplier Corning™ Supplier No. 4581

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid; Type: Optical Glass Base

High optical quality, glass bottom black microplates are ideal for performing high-content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.

  • High optical quality and scratch resistant glass
  • Glass bottom thickness of 200 μm is well suited for imaging microscopy.
  • Bottom flatness <50 μm to ensure planarity for imaging devices
  • Low background fluorescence and minimal cross-talk provide the highest possible optical quality for cell-based assays.

Specifications

Format Standard
Lid With Lid
Array 16 x 24
Color Black, Clear Bottom
Material Cycloolefin Copolymer
No. of Wells 384
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
For Use With (Application) For performing high content cell based assays using imaging systems
No. per Case 10/Cs.
No. per Pack 1/Pk.
Show More Show Less