missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 1536-Well Nontreated Cyclic Olefin Copolymer Plates

Catalog No. 12456703
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
12456703 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12456703 Supplier Corning™ Supplier No. 4564

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nonsterile microplates have smooth top and clear, flat bottom.

Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.

  • 127 μm film thickness
  • 536-well low base, black/clear or white/clear bottom microplates
  • Bar coded
  • Custom bar codes available for compatibility with the Kalypsys system and with UHTS systems
  • Low auto-fluorescence
  • Broad chemical resistance including DMSO and alcohol
  • High mechanical stability
  • Optimized for flatness and uniformity
  • Low birefringence

Specifications

Barcode Includes Barcode
Lid Without Lid
Array 32 x 48
Binding Type Poly-D-Lysine
Color Black, Clear Bottom
For Use With (Equipment) Automated dispensers
Material Cycloolefin Copolymer
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Poly-D-Lysine
Well Shape Flat Bottom (F-well)
For Use With (Application) For higher sensitivity
No. per Case 100/Cs.
No. per Pack 20/Pk.
Quantity 20/Pk.
Show More Show Less