Learn More
Corning™ 1536-Well, Multi-Treated, Flat-Bottom Microplate
Description
- No logo or lettering for improved heat sealing and ease of liquid handling
- Low base for optimal bottom reader sensitivity
- Signal dynamic range due to low background fluorescence and enhanced signal intensity
- Flatness minimizes variability within a plate and between plates
- Compatible with automated dispensers including the PerkinElmer FlexDrop™
- Recommended working volumes of up to 10μL
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Array | 32 x 48 |
| Color | Black, Clear Bottom |
| Material | Polystyrene |
| No. of Wells | 1536 |
| Surface Treatment | Multi-treated |
| Well Shape | Flat Bottom (F-well) |