Learn More
Corning™ 1536-Well, Non-Binding, Flat-Bottom, Bar-Coded Microplate
Description
Sold in bulk pack
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 32 x 48 |
| Color | White |
| No. of Wells | 1536 |
| Sterility | Non-sterile |
| Surface Treatment | Non-binding |
| Well Shape | Flat Bottom (F-well) |
| Quantity | 50/Cs. |