missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 1536-Well Black Clear-Bottom Polystyrene Plates

Catalog No. 0553930
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
0553930 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0553930 Supplier Corning™ Supplier No. 3891

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Highest density microplates available for high throughput screening

Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.

  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.

Order Info

Packaged in bags of 10

Specifications

Format Standard
Lid Without Lid
Array 32 x 48
Packaging 50/Pk.
Color Black, Clear Bottom
Product Type 1536-Well Assay Microplate
For Use With (Equipment) Automated dispensers
Material Polystyrene
Volume (Metric) Well 12.8 μL
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) For fluorescent homogeneous assays, high throughput screening
No. per Case 50/Cs.
No. per Pack 10/Pk.
Show More Show Less