missing translation for 'onlineSavingsMsg'
Learn More

Corn Starch, Powder, NF, Spectrum™ Chemical

Catalog No. p-7157843
Change view
Click to view available options
Quantity:
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601856 12 kg Poly Pail
18601857 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18601856 Supplier Spectrum Chemical Mfg Cor Supplier No. S155212KG
Only null left

S1552, 9005-25-8, (C6H10O5)n

Spectrum™ Chemical Corn Starch, Powder, NF is used as filler, binder and disintegrant excipient. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9005-25-8
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00082026,MFCD00132834
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
IUPAC Name 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

CAS Max % 100%
Quantity 12 kg
Grade NF
Ignition Residue 0.6%
Loss on Drying 15%
Packaging Poly Pail