Learn More
Eisco™ Cork Borer Sharpener
Features a superior, brass plated cone mounted to a wooden handle and hinged cutter blade set in slot
Supplier:  Eisco™ CH0324B
Catalog No. S80818
Only null left
Explore available promotions
Description
Description
  • Cork borer sharpener
  • Brass plated cone
  • Hinged cutter blade set in slot
  • Mounted on a wooden handle
  • Perfect for any science or home laboratory
  • For borers that are 4 to 27 mm in diameter
Specifications
Specifications
Cork Borers
Metal, Wood
Brown
Cork Borer Sharpener
Borers with a 4 to 27 mm Diameter
Product Suggestions
Product Suggestions