Learn More
Cork Borer Sets with Handles
Bore holes in corks and rubber stoppers.
Supplier:  Customer Channels Group (Ccg) H9663
 View more versions of this product
Catalog No. S50166C
Only null left
Description
Description
  • Consist of plated tubing with precision-ground cutting edges
  • Smooth plastic handles
  • Handles are stamped with a number designation
  • Punch included with each set
Specifications
Specifications
Cork Borer
4.8 to 16.7mm
Plated tubing, smooth Plastic handles
0.18 to 0.65 in.
To bore holes in corks and Rubber stoppers
9 Pack
Product Suggestions
Product Suggestions