Learn More
Cork Borer Sets with Handles
Bore holes in corks and rubber stoppers.
Supplier: Customer Channels Group (Ccg) H9664
Description
- Consist of plated tubing with precision-ground cutting edges
- Smooth plastic handles
- Handles are stamped with a number designation
- Punch included with each set
Chemical Identifiers
| Cork Borer | |
| To bore holes in corks and Rubber stoppers | |
| Plated tubing, smooth Plastic handles |
Specifications
| Cork Borer | |
| 4.8 to 20.2mm | |
| Plated tubing, smooth Plastic handles |
| 0.18 to 0.79 in. | |
| To bore holes in corks and Rubber stoppers | |
| 12 Pack |