missing translation for 'onlineSavingsMsg'
Learn More

(-)-Corey Lactone Benzoate 98.0+%, TCI America™

Catalog No. p-7125553
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2605200MG 200 mg
C26051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2605200MG Supplier TCI America Supplier No. C2605200MG
Only null left

Chemical Identifiers

CAS 39746-00-4
Molecular Formula C15H16O5
Molecular Weight (g/mol) 276.288
MDL Number MFCD00674102
InChI Key OBRRYUZUDKVCOO-FVCCEPFGSA-N
Synonym --corey lactone benzoate, corey lactone benzoate, 3ar,4s,5r,6as-4-hydroxymethyl-2-oxohexahydro-2h-cyclopenta b furan-5-yl benzoate, 3ar,4s,5r,6as-5-benzoyloxy hexahydro-4-hydroxymethyl-2h-cyclopenta b furan-2-one, 1s,5r,6s,7r-7-benzoyloxy-6-hydroxymethyl-2-oxabicyclo 3.3.0 octan-3-one, 3ar,4s,5r,6as-5-benzoyloxy hexahydro-4-hydroxymethyl-2h-cyclopentafuran-2-one, 3ar,4s,5r,6as-4-hydroxymethyl-2-oxo-3,3a,4,5,6,6a-hexahydrocyclopenta b furan-5-yl benzoate, 3ar,4s,5r,6as-4-hydroxymethyl-2-oxo-hexahydrocyclopenta b furan-5-yl benzoate, benzoyl-corey lactone, pubchem19127
PubChem CID 2724454
IUPAC Name [(3aR,4S,5R,6aS)-4-(hydroxymethyl)-2-oxo-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-5-yl] benzoate
SMILES C1C2C(CC(=O)O2)C(C1OC(=O)C3=CC=CC=C3)CO

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 200 mg
Formula Weight 276.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Corey Lactone Benzoate
EINECSNumber (9)-0284
TSCA No