missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corannulene 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 269°C |
| Quantity | 100 mg |
| Formula Weight | 250.30 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Corannulene |
Safety and Handling
| TSCA | No |