missing translation for 'onlineSavingsMsg'
Learn More

Corannulene 97.0+%, TCI America™

Catalog No. C257220MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C257220MG 20 mg
C2572100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C257220MG Supplier TCI America Supplier No. C257220MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5821-51-2
Molecular Formula C20H10
Molecular Weight (g/mol) 250.3
InChI Key VXRUJZQPKRBJKH-UHFFFAOYSA-N
Synonym Dibenzo[ghi,mno]fluoranthene
PubChem CID 11831840
SMILES C1=CC2=C3C4=C1C=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2

Specifications

Melting Point 269°C
Quantity 20 mg
Formula Weight 250.30
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Corannulene