missing translation for 'onlineSavingsMsg'
Learn More

Copper wire, 0.1mm (0.004in) dia, hard, Puratronic™, 99.9985% (metals basis)

Catalog No. AA10971H2
Change view
Click to view available options
Quantity:
10 m
50 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10971H2 50 m
AA10971G6 10 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA10971H2 Supplier Thermo Scientific Chemicals Supplier No. 010971H2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: approx. 0.07g/m | CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Appearance Copper colored wire
Density 8.94g/cm3
Quantity 50 m
Odor Odorless
Vapor Pressure 0 hPa
Form Wire
Assay Percent Range 99.9985% (Metals basis)
Chemical Name or Material Copper, Hard
EINECSNumber 231-159-6
RTECSNumber GL5325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only