missing translation for 'onlineSavingsMsg'
Learn More

Copper wire, 0.015mm (0.0006in) dia, 99.99% (metals basis)

Catalog No. AA43238G6
Change view
Click to view available options
Quantity:
10 m
50 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43238G6 10 m
AA43238H2 50 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43238G6 Supplier Thermo Scientific Chemicals Supplier No. 043238G6

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Appearance Copper colored wire
Quantity 10 m
Odor Odorless
Vapor Pressure 0 hPa
Form Wire
Assay Percent Range 99.99% (Metals basis)
Chemical Name or Material Copper
EINECSNumber 231-159-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only