missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Sulfate Solution, 1.6% with 0.6% Sulfuric Acid, Reagents
Copper Sulfate Solution, 1.6% with 0.6% Sulfuric Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$113.43 - $367.29
Specifications
| Boiling Point | 100°C |
|---|---|
| Chemical Name or Material | Copper Sulfate Solution, 1.6% with 0.6% Sulfuric Acid |
| CAS | 7732-18-5,7758-99-8 |
| Color | Blue |
| pH | <2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-335-565
|
Labchem Inc
CC2561004A |
4 L |
N/A
|
|
|||||
|
01-335-564
|
Labchem Inc
CC256100500A |
500 mL |
N/A
|
|
|||||
Specifications
| 100°C | |
| 7732-18-5,7758-99-8 | |
| <2 | |
| CuSO4/H2O/H2SO4 | |
| 1.6% | |
| Poly Bottle |
| Copper Sulfate Solution, 1.6% with 0.6% Sulfuric Acid | |
| Blue | |
| Liquid | |
| Copper(II) Sulfate | |
| Reagent | |
| 1.01 |