missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate, Pentahydrate, ACS, LabChem™

Catalog No. lc134051
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC134051 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. LC134051 missing translation for 'mfr' LabChem missing translation for 'supplierNo' LC134051
Only null left

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
CAS Max % 100
Physical Form Solid
Quantity 500 g
Linear Formula CuSO4·5H2O
UN Number UN3077
Solubility Information Soluble in water
Formula Weight 249.68
Percent Purity 98.0 to 102.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 2286kg/m3
Decomposition Information Fume; Sulfur compounds
Density 2286kg/m3
Chemical Name or Material Copper Sulfate, Pentahydrate
Show More Show Less
EINECSNumber 231-847-6
Recommended Storage Room Temperature