missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Sulfate, For Nitrogen (Nitrate), Certified, 2.0% (w/v) ±0.1%, LabChem™
Supplier: LabChem LC134204
Chemical Identifiers
| Clear, blue | |
| Poly Bottle | |
| Copper Sulfate |
| Liquid | |
| Room Temperature |
Specifications
| Clear, blue | |
| Poly Bottle | |
| Room Temperature |
| Liquid | |
| 4 L | |
| Copper Sulfate |